Product Details

SNP ID
rs138647485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35101263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTCTAGAACTGTGGCCCGTTTC[A/C]GCCGCTCTGCCTCCACCTGGAAGCC
Phenotype
MIM: 614730 MIM: 608292
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM214B PubMed Links

Gene Details

Gene
FAM214B
Gene Name
family with sequence similarity 214 member B
There are no transcripts associated with this gene.

Gene
PIGO
Gene Name
phosphatidylinositol glycan anchor biosynthesis class O
There are no transcripts associated with this gene.

Gene
STOML2
Gene Name
stomatin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287031.1 586 Missense Mutation NP_001273960.1
NM_001287032.1 586 Missense Mutation NP_001273961.1
NM_001287033.1 586 Missense Mutation NP_001273962.1
NM_013442.2 586 Missense Mutation NP_038470.1

View Full Product Details