Product Details

SNP ID
rs138915839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137228837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTCCGCAGTGCCGTCAGAGCAC[A/G]CCCACGCCTCGCGGAGGGGTGGCCA
Phenotype
MIM: 606073 MIM: 609826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYSRT1 PubMed Links

Gene Details

Gene
CYSRT1
Gene Name
cysteine rich tail 1
There are no transcripts associated with this gene.

Gene
NDOR1
Gene Name
NADPH dependent diflavin oxidoreductase 1
There are no transcripts associated with this gene.

Gene
RNF208
Gene Name
ring finger protein 208
There are no transcripts associated with this gene.

Gene
RNF224
Gene Name
ring finger protein 224
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190228.1 592 Silent Mutation ACA,ACG T74T NP_001177157.1
XM_011518914.2 592 Silent Mutation ACA,ACG T74T XP_011517216.1
Gene
SLC34A3
Gene Name
solute carrier family 34 member 3
There are no transcripts associated with this gene.

View Full Product Details