Product Details

SNP ID
rs139050031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130893705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACGGAGTGGGGTCGGGGCCCC[A/G]TGGCCAGGTCGGCCCAGCGTTCTCC
Phenotype
MIM: 189980 MIM: 613357 MIM: 609795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABL1 PubMed Links

Gene Details

Gene
ABL1
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
FIBCD1
Gene Name
fibrinogen C domain containing 1
There are no transcripts associated with this gene.

Gene
QRFP
Gene Name
pyroglutamylated RFamide peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198180.2 655 Missense Mutation ACG,ATG T45M NP_937823.1

View Full Product Details