Product Details

SNP ID
rs139277918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137030772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGCGGGGTAGGTGTGGGTAGC[A/G]GTCACAGATGGCACAGAAACGTTCC
Phenotype
MIM: 600047 MIM: 602030 MIM: 605798
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA2 PubMed Links

Gene Details

Gene
ABCA2
Gene Name
ATP binding cassette subfamily A member 2
There are no transcripts associated with this gene.

Gene
C9orf139
Gene Name
chromosome 9 open reading frame 139
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207511.2 1816 Intron NP_997394.1
XM_017014717.1 1816 Intron XP_016870206.1
Gene
FUT7
Gene Name
fucosyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004479.3 1816 Missense Mutation CGC,TGC R323C NP_004470.1
Gene
NPDC1
Gene Name
neural proliferation, differentiation and control 1
There are no transcripts associated with this gene.

View Full Product Details