Product Details

SNP ID
rs139301401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131434949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTGATCCAGCATTTCCATTTC[C/T]AAGGTCGTATGCTATGGAATTACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRRC2B PubMed Links

Gene Details

Gene
PRRC2B
Gene Name
proline rich coiled-coil 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013318.3 Intron NP_037450.2

View Full Product Details