Product Details

SNP ID
rs139546801
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137243747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCATTCCTTCCAGTAGGCCCTCC[A/G]GGTGCTGTTGCCGAATGTCAGGGCG
Phenotype
MIM: 609826 MIM: 602660
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C9orf173-AS1 PubMed Links
Additional Information
For this assay, SNP(s) [rs4073101] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C9orf173-AS1
Gene Name
C9orf173 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM166A
Gene Name
family with sequence similarity 166 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001710.2 1824 Missense Mutation CGG,TGG R303W NP_001001710.1
XM_006717108.2 1824 Missense Mutation CGG,TGG R287W XP_006717171.2
XM_011518694.1 1824 Missense Mutation CGG,TGG R338W XP_011516996.1
XM_011518695.1 1824 Missense Mutation CGG,TGG R300W XP_011516997.1
XM_011518696.2 1824 Intron XP_011516998.1
XM_011518697.1 1824 Intron XP_011516999.1
XM_017014718.1 1824 Missense Mutation CGG,TGG R216W XP_016870207.1
Gene
SLC34A3
Gene Name
solute carrier family 34 member 3
There are no transcripts associated with this gene.

Gene
STPG3
Gene Name
sperm-tail PG-rich repeat containing 3
There are no transcripts associated with this gene.

Gene
TUBB4B
Gene Name
tubulin beta 4B class IVb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006088.5 1824 Intron NP_006079.1

View Full Product Details