Product Details
- SNP ID
-
rs139562338
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:27524351 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAAAAAAAATGAGCACCAAACC[C/T]GATATGATTCAAAAGTGTTTGTGGC
- Phenotype
-
MIM: 615326
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IFNK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144060127,rs56087834] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFNK
- Gene Name
- interferon kappa
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020124.2 |
38 |
Silent Mutation |
CCC,CCT |
P5P |
NP_064509.2 |
- Gene
- MOB3B
- Gene Name
- MOB kinase activator 3B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024761.4 |
38 |
Intron |
|
|
NP_079037.3 |
View Full Product Details