Product Details

SNP ID
rs139563439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104569435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCCAGCTTTCTGGCAGTAAAG[A/G]AAAAGGTTTCCTTCTCTGGGTGTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR13C8 PubMed Links

Gene Details

Gene
OR13C8
Gene Name
olfactory receptor family 13 subfamily C member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004483.1 268 Missense Mutation AAA,GAA K90E NP_001004483.1

View Full Product Details