Product Details
- SNP ID
-
rs139657412
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21304885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGTCTTCCACTCCAACCTCCTG[A/C]ATCATACAGGCTTCCAGGTCATTCA
- Phenotype
-
MIM: 147565
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA5
PubMed Links
Gene Details
- Gene
- IFNA5
- Gene Name
- interferon alpha 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002169.2 |
429 |
Missense Mutation |
ATG,ATT |
M124I |
NP_002160.1 |
View Full Product Details