Product Details

SNP ID
rs139677853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83944962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTATCTGCTACAAGGATTTGGG[C/T]TCGTTTGTAAAAAGAAACTCTTTTC
Phenotype
MIM: 611342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf64 PubMed Links

Gene Details

Gene
C9orf64
Gene Name
chromosome 9 open reading frame 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317997.1 867 Missense Mutation ACC,GCC T68A NP_001304926.1
NM_032307.4 867 Missense Mutation ACC,GCC T209A NP_115683.3
XM_005252271.4 867 Intron XP_005252328.1

View Full Product Details