Product Details

SNP ID
rs140177554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127741876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCGGCCGCCAGCTGAATGGTC[C/T]TGTGCAGCAGCGCTGCCCGCTCCTC
Phenotype
MIM: 604722 MIM: 608052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH2D3C PubMed Links

Gene Details

Gene
SH2D3C
Gene Name
SH2 domain containing 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142531.1 1650 Missense Mutation NP_001136003.1
NM_001142532.1 1650 Missense Mutation NP_001136004.1
NM_001142533.1 1650 Missense Mutation NP_001136005.1
NM_001142534.1 1650 Missense Mutation NP_001136006.1
NM_001252334.1 1650 Missense Mutation NP_001239263.1
NM_005489.3 1650 Missense Mutation NP_005480.2
NM_170600.2 1650 Missense Mutation NP_733745.1
XM_005251639.1 1650 Missense Mutation XP_005251696.1
XM_011518114.1 1650 Missense Mutation XP_011516416.1
XM_011518115.1 1650 Missense Mutation XP_011516417.1
XM_011518117.2 1650 Missense Mutation XP_011516419.1
XM_017014174.1 1650 Missense Mutation XP_016869663.1
Gene
TOR2A
Gene Name
torsin family 2 member A
There are no transcripts associated with this gene.

View Full Product Details