Product Details

SNP ID
rs140199968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37780798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGGTTTTTGCCTTAACCCATA[C/T]TCTTCCATTCATTCCAAATACTATC
Phenotype
MIM: 606489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC3 PubMed Links

Gene Details

Gene
EXOSC3
Gene Name
exosome component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002269.2 757 UTR 3 NP_001002269.1
NM_016042.3 757 Missense Mutation NP_057126.2
Gene
LOC105376037
Gene Name
uncharacterized LOC105376037
There are no transcripts associated with this gene.

Gene
TRMT10B
Gene Name
tRNA methyltransferase 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286950.1 757 Intron NP_001273879.1
NM_001286951.1 757 Intron NP_001273880.1
NM_001286952.1 757 Intron NP_001273881.1
NM_001286953.1 757 Intron NP_001273882.1
NM_001286954.1 757 Intron NP_001273883.1
NM_144964.3 757 Intron NP_659401.2
XM_005251375.2 757 Intron XP_005251432.1
XM_005251379.2 757 Intron XP_005251436.1
XM_011517735.2 757 Intron XP_011516037.1
XM_011517736.2 757 Intron XP_011516038.1
XM_011517738.2 757 Intron XP_011516040.1
XM_011517739.2 757 Intron XP_011516041.1
XM_011517742.1 757 Intron XP_011516044.1
XM_011517743.2 757 Intron XP_011516045.1
XM_017014313.1 757 Intron XP_016869802.1

View Full Product Details