Product Details
- SNP ID
-
rs140281775
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:35957581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACAGGTGAAGTGGTTGATGACA[C/T]TGTCTCCACAGAAGGGCAGCTGAAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2S2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2233566] are located under a probe and SNP(s) [rs2233565] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2S2
- Gene Name
- olfactory receptor family 2 subfamily S member 2 (gene/pseudogene)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019897.2 |
574 |
Missense Mutation |
AAT,AGT |
N173S |
NP_063950.2 |
View Full Product Details