Product Details

SNP ID
rs140336222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108855075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGCCAATAGTGATGAGTTTGC[C/T]GTCCGGGAGCTCGTAGTCCACGCGC
Phenotype
MIM: 604303 MIM: 604304
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTL7A PubMed Links

Gene Details

Gene
ACTL7A
Gene Name
actin like 7A
There are no transcripts associated with this gene.

Gene
ACTL7B
Gene Name
actin like 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006686.3 921 Missense Mutation AGC,GGC S286G NP_006677.1

View Full Product Details