Product Details

SNP ID
rs140387263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137110921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTCCCCCCTGGATGGTGACGGC[A/G]ATGACTGAGGCACTCAGGTTCCTCC
Phenotype
MIM: 610537 MIM: 604346
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPP7 PubMed Links

Gene Details

Gene
DPP7
Gene Name
dipeptidyl peptidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013379.2 1332 Silent Mutation ATC,ATT I434I NP_037511.2
XM_005266075.3 1332 Silent Mutation ATC,ATT I507I XP_005266132.1
XM_006717083.3 1332 Missense Mutation CGC,TGC R485C XP_006717146.1
XM_011518599.2 1332 Silent Mutation ATC,ATT I456I XP_011516901.1
XM_011518600.1 1332 Silent Mutation ATC,ATT I426I XP_011516902.1
XM_017014651.1 1332 Missense Mutation CGC,TGC R434C XP_016870140.1
XM_017014652.1 1332 Missense Mutation CGC,TGC R412C XP_016870141.1
Gene
LOC101930307
Gene Name
uncharacterized LOC101930307
There are no transcripts associated with this gene.

Gene
MAN1B1
Gene Name
mannosidase alpha class 1B member 1
There are no transcripts associated with this gene.

View Full Product Details