Product Details
- SNP ID
-
rs140579362
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128733443 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCTTACCGGGCCATTTCCTTGGT[C/G]TCCTGCCGTGCGGTCGCCATCTTAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100506100
PubMed Links
Gene Details
- Gene
- LOC100506100
- Gene Name
- uncharacterized LOC100506100
There are no transcripts associated with this gene.
- Gene
- ZDHHC12
- Gene Name
- zinc finger DHHC-type containing 12
There are no transcripts associated with this gene.
- Gene
- ZER1
- Gene Name
- zyg-11 related cell cycle regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006336.3 |
2466 |
Missense Mutation |
GAC,GAG |
D742E |
NP_006327.2 |
XM_005251645.2 |
2466 |
Missense Mutation |
GAC,GAG |
D742E |
XP_005251702.1 |
XM_005251646.2 |
2466 |
Missense Mutation |
GAC,GAG |
D740E |
XP_005251703.1 |
XM_011518122.2 |
2466 |
Missense Mutation |
GAC,GAG |
D742E |
XP_011516424.1 |
XM_017014185.1 |
2466 |
Missense Mutation |
GAC,GAG |
D742E |
XP_016869674.1 |
XM_017014186.1 |
2466 |
Missense Mutation |
GAC,GAG |
D740E |
XP_016869675.1 |
XM_017014187.1 |
2466 |
Missense Mutation |
GAC,GAG |
D740E |
XP_016869676.1 |
XM_017014188.1 |
2466 |
Missense Mutation |
GAC,GAG |
D740E |
XP_016869677.1 |
XM_017014189.1 |
2466 |
Missense Mutation |
GAC,GAG |
D679E |
XP_016869678.1 |
XM_017014190.1 |
2466 |
Missense Mutation |
GAC,GAG |
D679E |
XP_016869679.1 |
View Full Product Details