Product Details

SNP ID
rs140628488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135501030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCGTCGCGCTGGTTGGGCTTT[C/G]TCGGGAAGGCGACCCCCCGGCCTGC
Phenotype
MIM: 614502 MIM: 611971
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C9orf116 PubMed Links

Gene Details

Gene
C9orf116
Gene Name
chromosome 9 open reading frame 116
There are no transcripts associated with this gene.

Gene
LOC101928525
Gene Name
uncharacterized LOC101928525
There are no transcripts associated with this gene.

Gene
MRPS2
Gene Name
mitochondrial ribosomal protein S2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016034.4 280 Missense Mutation CTC,GTC L26V NP_057118.1
XM_006717136.3 280 Missense Mutation CTC,GTC L49V XP_006717199.1

View Full Product Details