Product Details
- SNP ID
-
rs140800385
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:19118377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCAATGCTCCTTTTCCACTCTA[C/T]CCATGAGAGGTAGAGCTTATCCTGA
- Phenotype
-
MIM: 103195
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLIN2
PubMed Links
Gene Details
- Gene
- PLIN2
- Gene Name
- perilipin 2
View Full Product Details