Product Details
- SNP ID
-
rs140821304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:121212574 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGAATCCCCAGAGCATGGTATCTG[C/T]GACAAATTAAGCTAGACATAATTTT
- Phenotype
-
MIM: 137350
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSN
PubMed Links
Gene Details
- Gene
- GSN
- Gene Name
- gelsolin
View Full Product Details