Product Details

SNP ID
rs141012074
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27546767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTAGTATTTATATATTTTTTA[A/T]ATGACTGAGCTACAGTACAACAGTC
Phenotype
MIM: 614260
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
C9orf72 PubMed Links

Gene Details

Gene
C9orf72
Gene Name
chromosome 9 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256054.2 3022 UTR 3 NP_001242983.1
NM_018325.4 3022 UTR 3 NP_060795.1
NM_145005.6 3022 Intron NP_659442.2

View Full Product Details