Product Details

SNP ID
rs141139171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35650388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGATAATGCAGGTTCCCATACAG[C/T]GGGACCTCTTCCACAGACTCTCCAG
Phenotype
MIM: 107272 MIM: 157660 MIM: 604964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF39 PubMed Links

Gene Details

Gene
ARHGEF39
Gene Name
Rho guanine nucleotide exchange factor 39
There are no transcripts associated with this gene.

Gene
CCDC107
Gene Name
coiled-coil domain containing 107
There are no transcripts associated with this gene.

Gene
CD72
Gene Name
CD72 molecule
There are no transcripts associated with this gene.

Gene
LOC101926948
Gene Name
uncharacterized LOC101926948
There are no transcripts associated with this gene.

Gene
RMRP
Gene Name
RNA component of mitochondrial RNA processing endoribonuclease
There are no transcripts associated with this gene.

Gene
SIT1
Gene Name
signaling threshold regulating transmembrane adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014450.2 361 Silent Mutation CCA,CCG P88P NP_055265.1

View Full Product Details