Product Details

SNP ID
rs141247513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136946119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGAGGGGCTGTGCACGTGGTTGT[C/T]GCTGAGACCGACTACCAGAGTTTCG
Phenotype
MIM: 120930 MIM: 609072 MIM: 612905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8G PubMed Links

Gene Details

Gene
C8G
Gene Name
complement component 8, gamma polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000606.2 456 Silent Mutation GTC,GTT V127V NP_000597.2
Gene
FBXW5
Gene Name
F-box and WD repeat domain containing 5
There are no transcripts associated with this gene.

Gene
LCN12
Gene Name
lipocalin 12
There are no transcripts associated with this gene.

View Full Product Details