Product Details

SNP ID
rs141640684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129095562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTTATAGCAGACACCGTAGCC[A/G]TCGGGGACCACGGGCCCGAAGAACA
Phenotype
MIM: 600184 MIM: 614516
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRAT PubMed Links

Gene Details

Gene
CRAT
Gene Name
carnitine O-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000755.3 2097 Silent Mutation GAC,GAT D572D NP_000746.2
NM_001257363.1 2097 Silent Mutation GAC,GAT D551D NP_001244292.1
XM_005251708.3 2097 Silent Mutation GAC,GAT D551D XP_005251765.1
XM_017014274.1 2097 Silent Mutation GAC,GAT D573D XP_016869763.1
XM_017014275.1 2097 Silent Mutation GAC,GAT D573D XP_016869764.1
Gene
DOLPP1
Gene Name
dolichyldiphosphatase 1
There are no transcripts associated with this gene.

View Full Product Details