Product Details

SNP ID
rs141676121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:117704536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATGGCCTTCCTCTCCTGCGTG[A/G]GACCAGAAAGCTGGGAGCCCTGCGT
Phenotype
MIM: 603030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLR4 PubMed Links

Gene Details

Gene
TLR4
Gene Name
toll like receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003266.3 362 UTR 5 NP_003257.1
NM_138554.4 362 Missense Mutation AGA,GGA R22G NP_612564.1
NM_138557.2 362 UTR 5 NP_612567.1

View Full Product Details