Product Details
- SNP ID
-
rs141874070
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21141152 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGAAGTACCTCCTCAAGGTCAGT[A/G]CAGGGCTGCTAATTGCCCCAGCAGA
- Phenotype
-
MIM: 147553
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNW1
PubMed Links
Gene Details
- Gene
- IFNW1
- Gene Name
- interferon omega 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002177.2 |
750 |
Missense Mutation |
GCA,GTA |
A140V |
NP_002168.1 |
View Full Product Details