Product Details

SNP ID
rs141903692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128509036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGATCAACCCTCATTTGTTCCCA[A/G]ATCTCCTGACGCAAGCTCTGCCTTT
Phenotype
MIM: 603371 MIM: 602015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLE1 PubMed Links

Gene Details

Gene
GLE1
Gene Name
GLE1, RNA export mediator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003722.1 486 Missense Mutation AAA,AGA K87R NP_001003722.1
NM_001499.2 486 Missense Mutation AAA,AGA K87R NP_001490.1
XM_006717059.2 486 Missense Mutation AAA,AGA K90R XP_006717122.1
XM_006717060.2 486 Missense Mutation AAA,AGA K90R XP_006717123.1
XM_011518549.1 486 Missense Mutation AAA,AGA K90R XP_011516851.1
XM_011518550.1 486 Missense Mutation AAA,AGA K90R XP_011516852.1
XM_011518551.1 486 Missense Mutation AAA,AGA K87R XP_011516853.1
Gene
ODF2
Gene Name
outer dense fiber of sperm tails 2
There are no transcripts associated with this gene.

View Full Product Details