Product Details

SNP ID
rs141948750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:107285148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTAAAAGTACAGGAGATCTCA[A/G]TTATACATGAGAAACTTTTCAGTGT
Phenotype
MIM: 600062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD23B PubMed Links

Gene Details

Gene
RAD23B
Gene Name
RAD23 homolog B, nucleotide excision repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244713.1 Intron NP_001231642.1
NM_001244724.1 Intron NP_001231653.1
NM_002874.4 Intron NP_002865.1

View Full Product Details