Product Details

SNP ID
rs142125779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131525117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCAGGGTGGTCCCCTGGCTCAG[A/G]AAAGGTCCGCTTGTAGCTCCGCCCA
Phenotype
MIM: 607423 MIM: 609328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POMT1 PubMed Links

Gene Details

Gene
POMT1
Gene Name
protein O-mannosyltransferase 1
There are no transcripts associated with this gene.

Gene
UCK1
Gene Name
uridine-cytidine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135954.2 822 UTR 3 NP_001129426.1
NM_001261450.2 822 Missense Mutation CCT,TCT P244S NP_001248379.1
NM_001261451.2 822 Missense Mutation CCT,TCT P258S NP_001248380.1
NM_001318519.1 822 Missense Mutation CCT,TCT P230S NP_001305448.1
NM_031432.3 822 Missense Mutation CCT,TCT P253S NP_113620.1
XM_005272224.2 822 Missense Mutation CCT,TCT P267S XP_005272281.1

View Full Product Details