Product Details

SNP ID
rs142131032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124264258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGCTTCTCTGCCAGCTCGTCA[C/T]ACCCTAGACCCAGCTGTAGTCTGTG
Phenotype
MIM: 604884
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC613206 PubMed Links

Gene Details

Gene
LOC613206
Gene Name
myeloproliferative disease associated tumor antigen 5
There are no transcripts associated with this gene.

Gene
NEK6
Gene Name
NIMA related kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145001.2 Intron NP_001138473.1
NM_001166167.1 Intron NP_001159639.1
NM_001166168.1 Intron NP_001159640.1
NM_001166169.1 Intron NP_001159641.1
NM_001166170.1 Intron NP_001159642.1
NM_001166171.1 Intron NP_001159643.1
NM_014397.5 Intron NP_055212.2
XM_005251664.1 Intron XP_005251721.1
XM_006716936.2 Intron XP_006716999.1
XM_017014217.1 Intron XP_016869706.1

View Full Product Details