Product Details

SNP ID
rs142429302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37919926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGATGACTTCCGGGACACTGTC[A/G]AACGGAGGGCTGTTCTGACCCAGAA
Phenotype
MIM: 600314
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHB PubMed Links

Gene Details

Gene
SHB
Gene Name
SH2 domain containing adaptor protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003028.2 1993 Silent Mutation TTC,TTT F475F NP_003019.2

View Full Product Details