Product Details

SNP ID
rs142508708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5300354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGCTGGCTGCATCTCAGACAGG[G/T]TTAACTTCAGCTCCTGTGGCAAATT
Phenotype
MIM: 179730 MIM: 179740
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RLN1 PubMed Links

Gene Details

Gene
RLN1
Gene Name
relaxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006911.3 434 Intron NP_008842.1
Gene
RLN2
Gene Name
relaxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005059.3 434 UTR 3 NP_005050.2
NM_134441.2 434 Missense Mutation AAC,ACC N101T NP_604390.1
XM_017015011.1 434 Missense Mutation AAC,ACC N85T XP_016870500.1

View Full Product Details