Product Details
- SNP ID
-
rs143161006
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:74728322 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTGCATCTTCCCCCAAATTGGCC[A/G]GTCCAAACACCATTCCTCTTGATCT
- Phenotype
-
MIM: 607009
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRPM6
PubMed Links
Gene Details
- Gene
- TRPM6
- Gene Name
- transient receptor potential cation channel subfamily M member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001177310.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1946L |
NP_001170781.1 |
NM_001177311.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1946L |
NP_001170782.1 |
NM_017662.4 |
5814 |
Missense Mutation |
CCG,CTG |
P1951L |
NP_060132.3 |
XM_011518244.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1950L |
XP_011516546.1 |
XM_011518245.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1920L |
XP_011516547.1 |
XM_011518246.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1910L |
XP_011516548.1 |
XM_011518247.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1908L |
XP_011516549.1 |
XM_011518248.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1904L |
XP_011516550.1 |
XM_011518249.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1873L |
XP_011516551.1 |
XM_011518250.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1859L |
XP_011516552.1 |
XM_011518251.2 |
5814 |
Missense Mutation |
CCG,CTG |
P1708L |
XP_011516553.1 |
XM_011518252.2 |
5814 |
Intron |
|
|
XP_011516554.1 |
XM_011518255.2 |
5814 |
Intron |
|
|
XP_011516557.1 |
XM_017014287.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1830L |
XP_016869776.1 |
XM_017014288.1 |
5814 |
Missense Mutation |
CCG,CTG |
P1781L |
XP_016869777.1 |
XM_017014289.1 |
5814 |
Intron |
|
|
XP_016869778.1 |
View Full Product Details