Product Details

SNP ID
rs143226305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6534246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGTTTCCTCTCCTCTCCAATG[A/G]ATACAGGATTTACCATTCTCATTAT
Phenotype
MIM: 238300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLDC PubMed Links

Gene Details

Gene
GLDC
Gene Name
glycine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000170.2 Intron NP_000161.2

View Full Product Details