Product Details

SNP ID
rs143345996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135521569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGCCCACCACCTCCTGGCCTC[A/G]GACGAGGAGATTCAGGATGTGAGGC
Phenotype
MIM: 151675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCN1 PubMed Links

Gene Details

Gene
LCN1
Gene Name
lipocalin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252617.1 132 Silent Mutation TCA,TCG S24S NP_001239546.1
NM_001252618.1 132 Silent Mutation TCA,TCG S24S NP_001239547.1
NM_001252619.1 132 Silent Mutation TCA,TCG S24S NP_001239548.1
NM_002297.3 132 Silent Mutation TCA,TCG S24S NP_002288.1
XM_011518675.2 132 Silent Mutation TCA,TCG S88S XP_011516977.1

View Full Product Details