Product Details

SNP ID
rs143361281
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130893445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCCTTCACCGCCGACCGAAGC[A/G]GAAGCTGAAGCCGCCTTTCTTCCTG
Phenotype
MIM: 189980 MIM: 613357 MIM: 609795
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ABL1 PubMed Links

Gene Details

Gene
ABL1
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
FIBCD1
Gene Name
fibrinogen C domain containing 1
There are no transcripts associated with this gene.

Gene
QRFP
Gene Name
pyroglutamylated RFamide peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198180.2 915 Missense Mutation CGC,TGC R132C NP_937823.1

View Full Product Details