Product Details

SNP ID
rs143783247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35870156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCGGGATGACAGGAGGTGGAC[C/T]AGCATCAGAGGCACAAAGGTGGGCG
Phenotype
MIM: 616888
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00950 PubMed Links

Gene Details

Gene
LINC00950
Gene Name
long intergenic non-protein coding RNA 950
There are no transcripts associated with this gene.

Gene
OR13J1
Gene Name
olfactory receptor family 13 subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004487.1 246 Silent Mutation NP_001004487.1
Gene
TMEM8B
Gene Name
transmembrane protein 8B
There are no transcripts associated with this gene.

View Full Product Details