Product Details

SNP ID
rs143795003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:15422959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCAGTCTCCGGCAGTGGCGGCT[A/G]CAACTTTCCAGAGTATGAGCTTCCC
Phenotype
MIM: 602348
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNAPC3 PubMed Links

Gene Details

Gene
SNAPC3
Gene Name
small nuclear RNA activating complex polypeptide 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039697.1 264 Missense Mutation TAC,TGC Y27C NP_001034786.1
XM_017015056.1 264 Missense Mutation TAC,TGC Y27C XP_016870545.1
XM_017015057.1 264 Missense Mutation TAC,TGC Y27C XP_016870546.1

View Full Product Details