Product Details

SNP ID
rs144037445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137252504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAAAGCCCCTTCACGCAGAAAGC[C/T]GACTTCGACCAAGAGCAAAAGGTTG
Phenotype
MIM: 611180 MIM: 602660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf173-AS1 PubMed Links

Gene Details

Gene
C9orf173-AS1
Gene Name
C9orf173 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM166A
Gene Name
family with sequence similarity 166 member A
There are no transcripts associated with this gene.

Gene
NELFB
Gene Name
negative elongation factor complex member B
There are no transcripts associated with this gene.

Gene
STPG3
Gene Name
sperm-tail PG-rich repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004353.3 497 Silent Mutation GCC,GCT A157A NP_001004353.2
NM_001256699.1 497 Silent Mutation GCC,GCT A157A NP_001243628.1
NM_001256700.1 497 Silent Mutation GCC,GCT A157A NP_001243629.1
NM_001256701.1 497 Silent Mutation GCC,GCT A157A NP_001243630.1
XM_006717113.2 497 Silent Mutation GCC,GCT A157A XP_006717176.1
XM_006717115.2 497 Silent Mutation GCC,GCT A157A XP_006717178.1
Gene
TUBB4B
Gene Name
tubulin beta 4B class IVb
There are no transcripts associated with this gene.

View Full Product Details