Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033113.1 | 1268 | Missense Mutation | ATG,GTG | M475V | NP_001028285.1 |
NM_198585.2 | 1268 | Missense Mutation | ATG,GTG | M438V | NP_940987.2 |
XM_011518665.2 | 1268 | Missense Mutation | ATG,GTG | M583V | XP_011516967.1 |
XM_011518666.2 | 1268 | Missense Mutation | ATG,GTG | M546V | XP_011516968.1 |
XM_011518668.1 | 1268 | Missense Mutation | ATG,GTG | M348V | XP_011516970.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256067.1 | 1268 | Intron | NP_001242996.1 | ||
NM_001256068.1 | 1268 | Intron | NP_001242997.1 | ||
NM_006647.1 | 1268 | Intron | NP_006638.1 | ||
XM_011518158.2 | 1268 | Intron | XP_011516460.1 | ||
XM_011518159.1 | 1268 | Intron | XP_011516461.1 | ||
XM_011518160.1 | 1268 | Intron | XP_011516462.1 | ||
XM_017014220.1 | 1268 | Intron | XP_016869709.1 |