Product Details

SNP ID
rs144193548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136367684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGCCTGAGCCTGCCCTCCACGG[C/T]CAGTAGCTGCGCCTCACACTGGAAC
Phenotype
MIM: 607212 MIM: 609491 MIM: 602777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD9 PubMed Links

Gene Details

Gene
CARD9
Gene Name
caspase recruitment domain family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052813.4 1388 Missense Mutation ACC,GCC T408A NP_434700.2
NM_052814.3 1388 Missense Mutation ACC,GCC T408A NP_434701.1
Gene
DNLZ
Gene Name
DNL-type zinc finger
There are no transcripts associated with this gene.

Gene
GPSM1
Gene Name
G-protein signaling modulator 1
There are no transcripts associated with this gene.

Gene
SNAPC4
Gene Name
small nuclear RNA activating complex polypeptide 4
There are no transcripts associated with this gene.

View Full Product Details