Product Details

SNP ID
rs144229332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:89605736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACGAGGGCGACATCGCGCTGCA[A/G]ATCCATTTTACGCTGATCCAGGCTT
Phenotype
MIM: 604949
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GADD45G PubMed Links

Gene Details

Gene
GADD45G
Gene Name
growth arrest and DNA damage inducible gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006705.3 335 Silent Mutation CAA,CAG Q75Q NP_006696.1
XM_011518163.2 335 Silent Mutation CAA,CAG Q41Q XP_011516465.1

View Full Product Details