Product Details

SNP ID
rs144270600
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96641787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGGATAACTGAAGGTCTGTTT[A/G]TAAAGTTCACATGCTGAACTCCTAC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AAED1 PubMed Links

Gene Details

Gene
AAED1
Gene Name
AhpC/TSA antioxidant enzyme domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153698.1 689 Missense Mutation ACA,ATA T218I NP_714542.1
XM_005251783.3 689 Missense Mutation ACA,ATA T174I XP_005251840.1
XM_005251784.4 689 Intron XP_005251841.1

View Full Product Details