Product Details

SNP ID
rs144414973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:68419337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTCTCCATCACTGGCCATTTT[A/G]TGGGCAAAACAGCAAAACACAAATA
Phenotype
MIM: 600981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGM5 PubMed Links

Gene Details

Gene
PGM5
Gene Name
phosphoglucomutase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021965.3 Intron NP_068800.2
XM_011518782.2 Intron XP_011517084.1
XM_011518783.2 Intron XP_011517085.1
XM_017014808.1 Intron XP_016870297.1

View Full Product Details