Product Details

SNP ID
rs144714898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101362460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGGTCACATCTGGAATGAGGT[C/G]CTTCCTGAGAAATCTCTGGATCTCC
Phenotype
MIM: 602938
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BAAT PubMed Links

Gene Details

Gene
BAAT
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127610.1 1435 Missense Mutation CAC,GAC H409D NP_001121082.1
NM_001701.3 1435 Missense Mutation CAC,GAC H409D NP_001692.1

View Full Product Details