Product Details

SNP ID
rs144717868
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97658257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACCTTATTTCTACACTTCTTCT[C/T]TACCATGATTTGTAAACACCTTCTC
Phenotype
MIM: 600469 MIM: 611153
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NCBP1 PubMed Links

Gene Details

Gene
NCBP1
Gene Name
nuclear cap binding protein subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002486.4 Intron NP_002477.1
XM_011518715.2 Intron XP_011517017.1
XM_017014735.1 Intron XP_016870224.1
XM_017014736.1 Intron XP_016870225.1
XM_017014737.1 Intron XP_016870226.1
XM_017014738.1 Intron XP_016870227.1
Gene
XPA
Gene Name
XPA, DNA damage recognition and repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000380.3 Intron NP_000371.1
XM_006717278.1 Intron XP_006717341.1
XM_017015114.1 Intron XP_016870603.1

View Full Product Details