Product Details

SNP ID
rs144751431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:115894504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTTGTTCTAATTGGAATTTAT[G/T]GAACACTCCACTCAGCGGGAAACAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00474 PubMed Links

Gene Details

Gene
LINC00474
Gene Name
long intergenic non-protein coding RNA 474
There are no transcripts associated with this gene.

View Full Product Details