Product Details

SNP ID
rs145508640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35057132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACCACTTAGCCATACAGGTC[A/G]TCATCATTGTCTTCTGTGTATACAC
Phenotype
MIM: 601023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VCP PubMed Links

Gene Details

Gene
VCP
Gene Name
valosin containing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007126.3 2795 Silent Mutation GAC,GAT D802D NP_009057.1

View Full Product Details