Product Details

SNP ID
rs145821760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:3899941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAAGGGGTGGCAGCTTTAGATA[A/G]GAGATCAAGGAAAGCCTCTTGAAGA
Phenotype
MIM: 610192
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLIS3 PubMed Links

Gene Details

Gene
GLIS3
Gene Name
GLIS family zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042413.1 Intron NP_001035878.1
NM_152629.3 Intron NP_689842.3
XM_005251386.4 Intron XP_005251443.1
XM_005251387.4 Intron XP_005251444.1
XM_005251388.4 Intron XP_005251445.1
XM_005251389.4 Intron XP_005251446.1
XM_006716731.3 Intron XP_006716794.1
XM_011517763.2 Intron XP_011516065.1
XM_011517764.2 Intron XP_011516066.1
XM_011517765.2 Intron XP_011516067.1
XM_011517766.2 Intron XP_011516068.1
XM_011517767.2 Intron XP_011516069.1
XM_011517769.2 Intron XP_011516071.1
XM_017014361.1 Intron XP_016869850.1
Gene
GLIS3-AS1
Gene Name
GLIS3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details