Product Details

SNP ID
rs145905348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129042402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGCCCTGGCTGCCCACCAGCT[C/G]AAGAGGCGACGGAGGAGGAAGAAGC
Phenotype
MIM: 616774
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIGA2 PubMed Links

Gene Details

Gene
MIGA2
Gene Name
mitoguardin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005252282.1 400 Silent Mutation CTC,CTG L65L XP_005252339.1
XM_011519117.1 400 Silent Mutation CTC,CTG L129L XP_011517419.1
XM_011519118.1 400 UTR 5 XP_011517420.1

View Full Product Details