Product Details

SNP ID
rs146070656
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110199939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCTTAGGTTTAAAAATATATT[G/T]GATCCCAGGAAGGATCTGAGTGATC
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
C9orf152 PubMed Links

Gene Details

Gene
C9orf152
Gene Name
chromosome 9 open reading frame 152
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012993.2 2283 UTR 3 NP_001013011.2

View Full Product Details